accessibility menu, dialog, popup

UserName

NHN

CAS RN 2305-79-5

IUPAC Name: 4,5,6,7-tetrahydro-1H-indazole
Synonyms: 4,5,6,7-tetrahydro-1H-indazole
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: GDSQTWDUCDSZEY-UHFFFAOYSA-N
SMILES: C1CCC2=C(C1)NN=C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results