accessibility menu, dialog, popup

UserName

CAS RN 2303-01-7

CH3HOOSOOOHH3C

CAS RN 2303-01-7

IUPAC Name: 3,3-bis(4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: 3,3-bis(4-hydroxy-2-methylphenyl)-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 382.43
Molecular Formula: C21H18O5S
InChi Key: OLQIKGSZDTXODA-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)