accessibility menu, dialog, popup

UserName

CAS RN 22814-92-2

H2NNHSNHCl

CAS RN 22814-92-2

IUPAC Name: 3-amino-1-(4-chlorophenyl)thiourea
Synonyms: 3-amino-1-(4-chlorophenyl)thiourea
Molecular Weight (g/mol): 201.67
Molecular Formula: C7H8ClN3S
InChi Key: GFTWJLUVFRTLIL-UHFFFAOYSA-N
SMILES: NNC(=S)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results