CAS RN 22580-55-8
IUPAC Name:
pyrrolidin-2-one
Synonyms:
pyrrolidone 2-pyrol aminobutyric lactam 2-pyrrolidone-butyrolactam gamma-aminobutyric lactam 2-pyrrolidone, hydrotribromide gamma-aminobutyric acid lactam M-pyrol hydrobromide 2-pyrrolidone LAM
Molecular Weight (g/mol):
85.11
Molecular Formula:
C4H7NO
InChi Key:
HNJBEVLQSNELDL-UHFFFAOYSA-N
SMILES:
O=C1CCCN1
1
–
2
of
2
results
1
Pyrrolidone hydrotribromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pyrrolidone hydrotribromide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.