accessibility menu, dialog, popup

UserName

CAS RN 2217-41-6

NH2

CAS RN 2217-41-6

IUPAC Name: 5,6,7,8-tetrahydronaphthalen-1-amine
Synonyms: 5,6,7,8-tetrahydronaphthalen-1-amine
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: SODWJACROGQSMM-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=C1CCCC2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)