accessibility menu, dialog, popup

UserName

CAS RN 220141-73-1

H3COFFF

CAS RN 220141-73-1

IUPAC Name: 1-(3,4,5-trifluorophenyl)ethan-1-one
Synonyms: 1-(3,4,5-trifluorophenyl)ethanone
Molecular Weight (g/mol): 174.12
Molecular Formula: C8H5F3O
InChi Key: KXVIYAHOUAMVJX-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results