accessibility menu, dialog, popup

UserName

CAS RN 219861-08-2

H3CNCH3(S)ONFHOOOHO

CAS RN 219861-08-2

IUPAC Name: (1S)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile; oxalic acid
Synonyms: escitalopram; oxalic acid
Molecular Weight (g/mol): 414.43
Molecular Formula: C22H23FN2O5
InChi Key: KTGRHKOEFSJQNS-BDQAORGHSA-N
SMILES: OC(=O)C(O)=O.CN(C)CCC[C@]1(OCC2=CC(=CC=C12)C#N)C1=CC=C(F)C=C1
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results