accessibility menu, dialog, popup

UserName

CAS RN 2188-18-3

CH3CH3CH3OOHN(S)NH2NNHNOOOHO(E/Z)

CAS RN 2188-18-3

IUPAC Name: (2S)-5-{[amino(nitroamino)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: (2S)-5-{[amino(nitroamino)methylidene]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 319.32
Molecular Formula: C11H21N5O6
InChi Key: OZSSOVRIEPAIMP-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCCN=C(N)N[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)