accessibility menu, dialog, popup

UserName

CAS RN 218136-59-5

NNNNN(E/Z)

CAS RN 218136-59-5

IUPAC Name: 2,2-diphenyl-5-(4-{[(pyridin-3-yl)methylidene]amino}piperazin-1-yl)pentanenitrile
Synonyms: 2,2-diphenyl-5-{4-[(pyridin-3-ylmethylidene)amino]piperazin-1-yl}pentanenitrile
Molecular Weight (g/mol): 423.56
Molecular Formula: C27H29N5
InChi Key: QFYKXKMYVYOUNJ-UHFFFAOYSA-N
SMILES: N#CC(CCCN1CCN(CC1)N=CC1=CN=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results