accessibility menu, dialog, popup

UserName

CAS RN 2178-24-7

H3COOBr

CAS RN 2178-24-7

IUPAC Name: ethyl 2-(2-bromophenyl)acetate
Synonyms: ethyl 2-(2-bromophenyl)acetate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: MZQXAVZPEZUJIJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results