accessibility menu, dialog, popup

UserName

CH3HNNH2N

CAS RN 21703-52-6

IUPAC Name: 2-hydrazinyl-4-methylquinoline
Synonyms: 2-hydrazinyl-4-methylquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: FLDYBLAUDDQRAB-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC(NN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results