accessibility menu, dialog, popup

UserName

CAS RN 21508-19-0

OClO

CAS RN 21508-19-0

IUPAC Name: 5-chlorofuran-2-carbaldehyde
Synonyms: 5-chlorofuran-2-carbaldehyde
Molecular Weight (g/mol): 130.53
Molecular Formula: C5H3ClO2
InChi Key: DGAUAVDWXYXXGQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)