accessibility menu, dialog, popup

UserName

CAS RN 21436-44-2

CH3OOOCH3

CAS RN 21436-44-2

IUPAC Name: 3-methylbenzoyl 3-methylbenzoate
Synonyms: 3-methylbenzoyl 3-methylbenzoate
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: DGZSQLHPEZFGSI-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)C(=O)OC(=O)C1=CC(C)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results