accessibility menu, dialog, popup

UserName

CAS RN 214262-88-1

H3COOFCl

CAS RN 214262-88-1

IUPAC Name: methyl 2-(2-chloro-4-fluorophenyl)acetate
Synonyms: methyl 2-(2-chloro-4-fluorophenyl)acetate
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: DFTYYBRLWCAUHP-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=C(F)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results