CAS RN 213203-65-7
IUPAC Name:
4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene
Synonyms:
4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol):
257.03
Molecular Formula:
C8H5BrF4
InChi Key:
MOVSRIIBGRAUAF-UHFFFAOYSA-N
SMILES:
FC1=CC(CBr)=CC=C1C(F)(F)F
1
–
1
of
1
results
1
3-Fluoro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
CAS: 213203-65-7 Molecular Formula: C8H5BrF4 Molecular Weight (g/mol): 257.03 MDL Number: MFCD00236289 InChI Key: MOVSRIIBGRAUAF-UHFFFAOYSA-N Synonym: 3-fluoro-4-trifluoromethyl benzyl bromide,4-bromomethyl-2-fluoro-1-trifluoromethyl benzene,4-bromomethyl-2-fluoro-1-trifluoromethyl-benzene,acmc-20ai27,3-fluoro-4-trifluoromethylbenzyl bromide,3-fluoro-4-trifluoromethyl benzylbromide,3-fluoro-4-trifluoromethyl-benzyl bromide,benzene, 4-bromomethyl-2-fluoro-1-trifluoromethyl,benzene,4-bromomethyl-2-fluoro-1-trifluoromethyl PubChem CID: 2737574 IUPAC Name: 4-(bromomethyl)-2-fluoro-1-(trifluoromethyl)benzene SMILES: FC1=CC(CBr)=CC=C1C(F)(F)F