accessibility menu, dialog, popup

UserName

CAS RN 209803-40-7

CH3H3CNH2NCH3H3C

CAS RN 209803-40-7

IUPAC Name: (5-aminopentyl)bis(propan-2-yl)amine
Synonyms: (5-aminopentyl)diisopropylamine
Molecular Weight (g/mol): 186.34
Molecular Formula: C11H26N2
InChi Key: CIFBCEXYFFKMEB-UHFFFAOYSA-N
SMILES: CC(C)N(CCCCCN)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results