CAS RN 2064175-41-1
CAS RN 2064175-41-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ dTAG-13
Degrades mutant FKBP12F36V fusion proteins; useful alternative to genetic methods for target validation