accessibility menu, dialog, popup

UserName

CAS RN 20596-34-3

PtBrBrBrBrBrH2OH2OH2OH2OH2OH2OH2OH2OH2OBrH3OH3O

CAS RN 20596-34-3

IUPAC Name: platinum(4+) dihydrogen hexabromide
Synonyms: platinum(4+) dihydrogen hexabromide
Molecular Weight (g/mol): 676.52
Molecular Formula: Br6H2Pt
InChi Key: VOEHEFXBCNCSCK-UHFFFAOYSA-J
SMILES: [H+].[H+].[Br-].[Br-].[Br-].[Br-].[Br-].[Br-].[Pt+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results