CAS RN 204005-46-9
CAS RN 204005-46-9
IUPAC Name:
(3Z)-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylidene]-2,3-dihydro-1H-indol-2-one
Synonyms:
semaxanib
Molecular Weight (g/mol):
238.29
Molecular Formula:
C15H14N2O
InChi Key:
WUWDLXZGHZSWQZ-WQLSENKSSA-N
SMILES:
CC1=CC(C)=C(N1)\C=C1/C(=O)NC2=CC=CC=C12
1
–
2
of
2
results
1
Tocris Bioscience™ SU 5416
CAS: 204005-46-9 Molecular Formula: C15H14N2O
2
STEMCELL Technologies SU5416
Tyrosine kinase inhibitor; Inhibits KDR, PDGFR, KIT, RET, FLT-3, ABL, and ALK