accessibility menu, dialog, popup

UserName

CAS RN 203302-97-0

IUPAC Name: 2-[3-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [3-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: NFZQVADYFXRRPM-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results