accessibility menu, dialog, popup

UserName

CAS RN 202409-33-4

CH3NNClSH3COO

CAS RN 202409-33-4

IUPAC Name: 5-chloro-3-(4-methanesulfonylphenyl)-6'-methyl-2,3'-bipyridine
Synonyms: etoricoxib arcoxia nucoxia tauxib algix etoricoxibe C18H15clN2O2S etoricoxib (product) etoricoxib (substance) merck sharp & dohme brand of etoricoxib
Molecular Weight (g/mol): 358.84
Molecular Formula: C18H15ClN2O2S
InChi Key: MNJVRJDLRVPLFE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=N1)C1=NC=C(Cl)C=C1C1=CC=C(C=C1)S(C)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)