accessibility menu, dialog, popup

UserName

H3COOHH3CH3CCH3CH3H3CCH3

CAS RN 1991-52-2

IUPAC Name: 2,5-di-tert-butyl-4-methoxyphenol
Synonyms: 2,5-di-tert-butyl-4-methoxyphenol
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: FLLRQABPKFCXSO-UHFFFAOYSA-N
SMILES: COC1=CC(=C(O)C=C1C(C)(C)C)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results