accessibility menu, dialog, popup

UserName

BrNCS

CAS RN 1985-12-2

IUPAC Name: 1-bromo-4-isothiocyanatobenzene
Synonyms: P-bromophenyl isothiocyanate
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: XQACWEBGSZBLRG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)