accessibility menu, dialog, popup

UserName

CAS RN 19719-28-9

OHOClCl

CAS RN 19719-28-9

IUPAC Name: 2-(2,4-dichlorophenyl)acetic acid
Synonyms: (2,4-dichlorophenyl)acetic acid
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: GXMWLJKTGBZMBH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (10)
  • (1)
Concentration 
  • (4)
  • (1)
  • (2)
Solvent or Matrix 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (10)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)