CAS RN 19567-78-3
CAS RN 19567-78-3
IUPAC Name:
disodium hexachloroiridiumdiuide hexahydrate
Synonyms:
disodium hexachloroiridate(2-) hexahydrate
Molecular Weight (g/mol):
558.99
Molecular Formula:
Cl6H12IrNa2O6
InChi Key:
QZVJPVJIAOOTJU-UHFFFAOYSA-H
SMILES:
O.O.O.O.O.O.[Na+].[Na+].Cl[Ir--](Cl)(Cl)(Cl)(Cl)Cl
1
–
2
of
2
results
1
Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min
CAS: 19567-78-3 Molecular Formula: Cl6H12IrNa2O6 Molecular Weight (g/mol): 558.987 MDL Number: MFCD00149169 InChI Key: QZVJPVJIAOOTJU-UHFFFAOYSA-H Synonym: sodium hexachloroiridate iv hexahydrate,cl6ir.2na.6h2o,sodium hexachloroiridate 2--water 2/1/6,disodium hexachloroiridate 2-hexahydrate PubChem CID: 19849276 IUPAC Name: disodium;hexachloroiridium(2-);hexahydrate SMILES: O.O.O.O.O.O.[Na+].[Na+].Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl