CAS RN 194019-11-9
IUPAC Name:
2-(3-bromo-4-fluorophenyl)acetic acid
Synonyms:
(3-bromo-4-fluorophenyl)acetic acid
Molecular Weight (g/mol):
233.04
Molecular Formula:
C8H6BrFO2
InChi Key:
XXFGIJYSXNXNAU-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(Br)=C(F)C=C1
1
–
1
of
1
results
1
3-Bromo-4-fluorophenylacetic acid, 96%, Thermo Scientific™
CAS: 194019-11-9 Molecular Formula: C8H6BrFO2 Molecular Weight (g/mol): 233.04 MDL Number: MFCD04038373 InChI Key: XXFGIJYSXNXNAU-UHFFFAOYSA-N Synonym: 3-bromo-4-fluorophenylacetic acid,2-3-bromo-4-fluorophenyl acetic acid,3-bromo-4-fluorophenylaceticacid,3-bromo-4-fluorophenyl acetic acid,benzeneacetic acid, 3-bromo-4-fluoro,3-bromo-4-fluoro-phenyl-acetic acid,2-3-bromo-4-fluoro-phenyl acetic acid,acmc-209exn,ksc497i4b,61150-59-2 3-bromo-4-fluorophenylacetic acid PubChem CID: 3416339 IUPAC Name: 2-(3-bromo-4-fluorophenyl)acetic acid SMILES: OC(=O)CC1=CC(Br)=C(F)C=C1