CAS RN 19342-73-5
CAS RN 19342-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Deazariboflavin, TRC
CAS: 19342-73-5 Molecular Formula: C18 H21 N3 O6 Molecular Weight (g/mol): 375.38 Synonym: D-Ribitol, 1-deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxopyrimido[4,5-b]quinolin-10(2H)-yl)-,Pyrimido[4,5-b]quinoline-2,4(3H,10H)-dione, 7,8-dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)- (8CI),1-Deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxopyrimido[4,5-b]quinolin-10(2H)-yl)-D-ribitol,Pyrimido[4,5-b]quinoline, D-ribitol deriv.,5-Deazariboflavin,5-Deazariboflavine,NCI 0112202,NSC 112202 IUPAC Name: 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]pyrimido[4,5-b]quinoline-2,4-dione SMILES: Cc1cc2C=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C