CAS RN 193013-76-2
CAS RN 193013-76-2
IUPAC Name:
4'-(bromomethyl)-2-fluoro-1,1'-biphenyl
Synonyms:
4'-(bromomethyl)-2-fluoro-1,1'-biphenyl
Molecular Weight (g/mol):
265.13
Molecular Formula:
C13H10BrF
InChi Key:
SAUALJYDYRRWPW-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1C1=CC=C(CBr)C=C1
1
–
1
of
1
results
1
4-Bromomethyl-2'-fluorobiphenyl, 97%
CAS: 193013-76-2 Molecular Formula: C13H10BrF Molecular Weight (g/mol): 265.125 MDL Number: MFCD15146811 InChI Key: SAUALJYDYRRWPW-UHFFFAOYSA-N Synonym: 4'-bromomethyl-2-fluoro-1,1'-biphenyl,4-bromomethyl-2'-fluorobiphenyl,2-fluoro-4'-bromomethylbiphenyl PubChem CID: 18680932 IUPAC Name: 1-(bromomethyl)-4-(2-fluorophenyl)benzene SMILES: C1=CC=C(C(=C1)C2=CC=C(C=C2)CBr)F