accessibility menu, dialog, popup

UserName

CAS RN 192863-35-7

BFFFFK

CAS RN 192863-35-7

IUPAC Name: potassium trifluoro(4-fluorophenyl)boranuide
Synonyms: potassium trifluoro(4-fluorophenyl)boranuide
Molecular Weight (g/mol): 202
Molecular Formula: C6H4BF4K
InChi Key: PXKNUQDMYBUYSZ-UHFFFAOYSA-N
SMILES: [K+].FC1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results