accessibility menu, dialog, popup

UserName

CAS RN 1927-25-9

NH2(S)HOOHO

CAS RN 1927-25-9

IUPAC Name: (2S)-2-amino-4-hydroxybutanoic acid
Synonyms: L-homoserine L-of homoserine homoserine L- L-isomer of homoserine L isomer of homoserine homoserine L-isomer
Molecular Weight (g/mol): 119.12
Molecular Formula: C4H9NO3
InChi Key: UKAUYVFTDYCKQA-VKHMYHEASA-N
SMILES: N[C@@H](CCO)C(O)=O
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)