accessibility menu, dialog, popup

UserName

CAS RN 191-24-2

CAS RN 191-24-2

IUPAC Name: hexacyclo[12.8.0.0²,¹¹.0³,⁸.0⁴,²¹.0¹⁷,²²]docosa-1(22),2,4,6,8,10,12,14,16,18,20-undecaene
Synonyms: benzo(ghi)perylene 1,12-benzoperylene benzo(ghi)pyrilene ccris 784 1,12-benzperylene
Molecular Weight (g/mol): 276.34
Molecular Formula: C22H12
InChi Key: GYFAGKUZYNFMBN-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C3C=CC4=CC=C5C=CC=C6C(=C1)C2=C3C4=C56
Molecular Weight (g/mol) 
  • (6)
  • (7)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (7)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)