accessibility menu, dialog, popup

UserName

H3COCH3NCS

CAS RN 190774-56-2

IUPAC Name: 2-isothiocyanato-1-methoxy-4-methylbenzene
Synonyms: 2-isothiocyanato-1-methoxy-4-methylbenzene
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: CYKFAZDMLSXWKZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1N=C=S
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)