accessibility menu, dialog, popup

UserName

CAS RN 18911-76-7

OOOOOOCCCHHHYCF3CF3CF3CF3CF3CF3

CAS RN 18911-76-7

IUPAC Name: tris(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) yttrium
Synonyms: tris(1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one) yttrium
Molecular Weight (g/mol): 713.08
Molecular Formula: C15H6F18O6Y
InChi Key: QYKMGZLZIBNPQL-UHFFFAOYSA-N
SMILES: [Y].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results