accessibility menu, dialog, popup

UserName

CAS RN 18880-04-1

BrClCl

CAS RN 18880-04-1

IUPAC Name: 4-(bromomethyl)-1,2-dichlorobenzene
Synonyms: 4-(bromomethyl)-1,2-dichlorobenzene
Molecular Weight (g/mol): 239.92
Molecular Formula: C7H5BrCl2
InChi Key: XLWSBDFQAJXCQX-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)