accessibility menu, dialog, popup

UserName

CAS RN 185391-33-7

HNOONHF

CAS RN 185391-33-7

IUPAC Name: N-[(4-fluorophenyl)methyl]-2-(1H-indol-3-yl)-2-oxoacetamide
Synonyms: N-[(4-fluorophenyl)methyl]-2-(1H-indol-3-yl)-2-oxoacetamide
Molecular Weight (g/mol): 296.3
Molecular Formula: C17H13FN2O2
InChi Key: VWCCHJFFYCGXFL-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC(=O)C(=O)C2=CNC3=CC=CC=C23)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results