accessibility menu, dialog, popup

UserName

CAS RN 185331-69-5

NOOFBr

CAS RN 185331-69-5

IUPAC Name: 1-bromo-2-fluoro-4-nitrobenzene
Synonyms: 1-bromo-2-fluoro-4-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: AZYQMHQOHNGPRR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results