CAS RN 184901-82-4
CAS RN 184901-82-4
IUPAC Name:
2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Synonyms:
2-amino-N-{1-[(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamoyl]ethyl}propanamide; trifluoroacetic acid
Molecular Weight (g/mol):
453.84
Molecular Formula:
C18H23ClF3N3O5
InChi Key:
ZFZXFWAYCYJFIZ-UHFFFAOYNA-N
SMILES:
OC(=O)C(F)(F)F.CC(N)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)CCl
1
–
1
of
1
results
1
Thermo Scientific Chemicals Tripeptidyl Peptidase Inhibitor
CAS: 184901-82-4 Molecular Formula: C16H22ClN3O3,C18H23ClF3N3O5 Synonym: H-AAF-CMK; TPPII Inhibitor