accessibility menu, dialog, popup

UserName

CH3OHOFF

CAS RN 183237-86-7

IUPAC Name: 4,5-difluoro-2-methylbenzoic acid
Synonyms: 4,5-difluoro-2-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: LDMHYYGJZWDBQL-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(F)C=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results