CAS RN 182815-44-7
CAS RN 182815-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Colesevelam Hydrochloride (Technical Grade), TRC
CAS: 182815-44-7 Molecular Formula: (C12H27ClN2 • C13H27N • C3H7N • C3H5ClO)x • (HCl)x Molecular Weight (g/mol): 618.24 Synonym: CholestaGel,GT 31-104,GT 31-104HB,Welchol; IUPAC Name: 1-Hexanaminium, N,N,N-trimethyl-6-(2-propen-1-ylamino)-, chloride (1:1), polymer with 2-(chloromethyl)oxirane, 2-propen-1-amine and N-2-propen-1-yl-1-decanamine, hydrochloride (ACI) SMILES: C=CCNCCCCCC[N+](C)(C)C.C=CCNCCCCCCCCCC.C=CCN.ClCC1CO1.[Cl-].[HCl]