accessibility menu, dialog, popup

UserName

CAS RN 1820954-21-9

(S)(S)HNN(S)(S)(R)OOHOHCH3SNHSNH2OOH(R)CH3HO

CAS RN 1820954-21-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results