accessibility menu, dialog, popup

UserName

CAS RN 18144-43-9

H3CCH3OONH2

CAS RN 18144-43-9

IUPAC Name: propan-2-yl 4-aminobenzoate
Synonyms: isopropyl 4-aminobenzoate
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: JWCPZKNBPMSYND-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results