accessibility menu, dialog, popup

UserName

CAS RN 17609-47-1

CH3OO(S)NH2H3CCH3HCl

CAS RN 17609-47-1

IUPAC Name: hydrogen ethyl (2S)-2-amino-3-methylbutanoate chloride
Synonyms: hydrogen ethyl (2S)-2-amino-3-methylbutanoate chloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: PQGVTLQEKCJXKF-RGMNGODLSA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@@H](N)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)