CAS RN 175202-34-3
CAS RN 175202-34-3
IUPAC Name:
2-(thiophene-2-sulfonyl)ethanethioamide
Synonyms:
2-(thiophene-2-sulfonyl)ethanethioamide
Molecular Weight (g/mol):
221.31
Molecular Formula:
C6H7NO2S3
InChi Key:
YNPIRUTXXLFXRU-UHFFFAOYSA-N
SMILES:
NC(=S)CS(=O)(=O)C1=CC=CS1
1
–
1
of
1
results
1
2-(2-Thienylsulfonyl)ethanethioamide, 97%, Thermo Scientific™
CAS: 175202-34-3 Molecular Formula: C6H7NO2S3 Molecular Weight (g/mol): 221.307 MDL Number: MFCD00052277 InChI Key: YNPIRUTXXLFXRU-UHFFFAOYSA-N Synonym: 2-2-thienylsulfonyl ethanethioamide,2-thiophene-2-sulfonyl ethanethioamide,ethanethioamide,2-2-thienylsulfonyl,maybridge1_004617,2-2-thienyl sulfonyl thioacetamide,2-thiophen-2-ylsulfonyl ethanethioamide,2-thien-2-yl sulphonyl ethanethioamide,2-2-amino-2-thioxoethyl sulphonyl thiophene,2-amino-1-2-thienylsulfonyl ethane-2-thione PubChem CID: 2778709 IUPAC Name: 2-thiophen-2-ylsulfonylethanethioamide SMILES: C1=CSC(=C1)S(=O)(=O)CC(=S)N