accessibility menu, dialog, popup

UserName

CAS RN 175137-02-7

CH3OSClNH2

CAS RN 175137-02-7

IUPAC Name: 1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethan-1-one
Synonyms: 1-[3-amino-5-(4-chlorophenyl)thiophen-2-yl]ethanone
Molecular Weight (g/mol): 251.73
Molecular Formula: C12H10ClNOS
InChi Key: DLQVOIFYJQATMI-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(N)C=C(S1)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results