accessibility menu, dialog, popup

UserName

CAS RN 17440-83-4

HH2OH2OClH2NNH2NONClNH2NH2N

CAS RN 17440-83-4

IUPAC Name: hydrogen 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide dihydrate chloride
Synonyms: hydrogen amiloride dihydrate chloride
Molecular Weight (g/mol): 302.12
Molecular Formula: C6H13Cl2N7O3
InChi Key: LTKVFMLMEYCWMK-UHFFFAOYSA-N
SMILES: [H+].O.O.[Cl-].NC(N)=NC(=O)C1=NC(Cl)=C(N)N=C1N
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)