accessibility menu, dialog, popup

UserName

CAS RN 17397-89-6

CH3O(S)O(R)NH2O(E)(E)

CAS RN 17397-89-6

IUPAC Name: (2R,3S)-3-[(4E,7E)-nona-4,7-dienoyl]oxirane-2-carboxamide
Synonyms: cerulenin helicocerin
Molecular Weight (g/mol): 223.27
Molecular Formula: C12H17NO3
InChi Key: GVEZIHKRYBHEFX-NQQPLRFYSA-N
SMILES: C\C=C\C\C=C\CCC(=O)[C@H]1O[C@H]1C(N)=O
Color 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)