accessibility menu, dialog, popup

UserName

CAS RN 1737-28-6

H3COFFFOH

CAS RN 1737-28-6

IUPAC Name: 1-[4-(trifluoromethoxy)phenyl]ethan-1-ol
Synonyms: 1-[4-(trifluoromethoxy)phenyl]ethanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: RFESEZYNEIOJHS-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results