accessibility menu, dialog, popup

UserName

CH3NNBrNH2

CAS RN 17321-93-6

IUPAC Name: 5-bromo-4-methylpyrimidin-2-amine
Synonyms: 5-bromo-4-methylpyrimidin-2-amine
Molecular Weight (g/mol): 188.03
Molecular Formula: C5H6BrN3
InChi Key: LPQVTZJEIXYDQA-UHFFFAOYSA-N
SMILES: CC1=NC(N)=NC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results