CAS RN 173039-10-6
CAS RN 173039-10-6
IUPAC Name:
2-(phosphonomethyl)pentanedioic acid
Synonyms:
2-(phosphonomethyl)pentanedioic acid
Molecular Weight (g/mol):
226.12
Molecular Formula:
C6H11O7P
InChi Key:
ISEYJGQFXSTPMQ-UHFFFAOYNA-N
SMILES:
OC(=O)CCC(CP(O)(O)=O)C(O)=O
1
–
1
of
1
results
1
Tocris Bioscience™ PMPA (NAALADase inhibitor)
CAS: 173039-10-6 Molecular Formula: C6H11O7P