CAS RN 1726-23-4
CAS RN 1726-23-4
IUPAC Name:
1,2,4-tributyl benzene-1,2,4-tricarboxylate
Synonyms:
1,2,4-tributyl benzene-1,2,4-tricarboxylate
Molecular Weight (g/mol):
378.47
Molecular Formula:
C21H30O6
InChi Key:
RJIFVNWOLLIBJV-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C1=CC=C(C(=O)OCCCC)C(=C1)C(=O)OCCCC
1
–
3
of
3
results
1
Tributyl Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tributyl Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tributyl Trimellitate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.